Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Plays a role in the negative regulation of NF-kappa-B signaling at the basal level by modulating transcriptional activity of NF-kappa-B on its target gene promoters. Associates with cyclin D1 promoter containing Myc E-box sequence and transcriptionally represses cyclin D1 expression. Regulates telomerase reverse transcriptase expression and telomerase activity in both ALT (alternative lengthening of telomeres)and telomerase-positive cell lines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TCEA-like protein 7; transcription elongation factor A (SII)-like 7; Transcription elongation factor A protein-like 7; Transcription elongation factor S-II protein-like 7; unnamed protein product
Gene Aliases: TCEAL7; WEX5
UniProt ID: (Human) Q9BRU2
Entrez Gene ID: (Human) 56849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support