Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: rRNA-processing protein UTP23 homolog; unnamed protein product; UTP23, small subunit (SSU) processome component, homolog
Gene Aliases: 1700010I21Rik; AI662478; BB238373; C8orf53; D530033C11Rik; UTP23
UniProt ID: (Human) Q9BRU9, (Mouse) Q9CX11
Entrez Gene ID: (Human) 84294, (Mouse) 78581
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support