Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fibronectin type III, immunoglobulin and leucine rich repeat domains 5; Leucine-rich repeat neuronal protein 3; leucine-rich repeat protein, neuronal 3; LRP; LRRN3; Neuronal leucine-rich repeat protein 3; NLRR-3; unnamed protein product
Gene Aliases: FIGLER5; LRRN3; Nbla10363; NLRR-3; NLRR3; UNQ194/PRO220
UniProt ID: (Human) Q9H3W5
Entrez Gene ID: (Human) 54674
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support