Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Actin-like protein 8 (ACTL8), also known as Cancer/Testis Antigen 57 (CT57), is part of the cancer/testis antigen (CTA) family. It is primarily expressed in the testes but aberrantly overexpressed in various cancers, including lung adenocarcinoma, endometrial cancer, and colorectal cancer. ACTL8's overexpression is associated with enhanced cell mobility, proliferation, and colony formation, contributing to tumor growth and poor prognosis. The gene is believed to play a critical role in oncogenesis and is being investigated as a potential therapeutic target due to its involvement in tumorigenic processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin like protein; Actin-like protein 8; Cancer/testis antigen 57; CT57; peptide with resemblance to the actin family; the actual start of the coding region has not been determined; unnamed protein product
Gene Aliases: ACTL8; CT57
UniProt ID: (Human) Q9H568
Entrez Gene ID: (Human) 81569
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support