Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LQLEREMALA TNRSLAERNL EFQGPLEISR SNLSDRYQEL RKLVERCQEQ
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
VPS37C is a subunit of ESCRT-I pathway required for sorting ubiquitinated transmembrane proteins into internal vesicles of multivesicular bodies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ESCRT-I complex subunit VPS37C; FLJ20847; hVps37C; unnamed protein product; vacuolar protein sorting 37 homolog C; vacuolar protein sorting 37C; Vacuolar protein sorting-associated protein 37C; VPS37C, ESCRT-I subunit
Gene Aliases: PML39; VPS37C
UniProt ID: (Human) A5D8V6
Entrez Gene ID: (Human) 55048
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support