Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NLIHEPLDQG NCAGSWAFST AAVASDRVSI HSLGHMTPVL SPQNLLSCDT
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: androgen-regulated gene 1; Glucocorticoid-inducible protein 5; lipocalin 7; OLRG-2; Oxidized LDL-responsive gene 2 protein; oxidized-LDL responsive gene 2; P3ECSL; TIN Ag-related protein; TIN-ag; TIN-Ag-RP; TINAG-like 1; Tubulointerstitial nephritis antigen-like; Tubulointerstitial nephritis antigen-related protein; unnamed protein product
Gene Aliases: ARG1; GIS5; LCN7; LIECG3; OLRG2; PP6614; PSEC0088; TINAGL; TINAGL1; TINAGRP; UNQ204/PRO230
UniProt ID: (Human) Q9GZM7
Entrez Gene ID: (Human) 64129
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support