Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • CITED2 Antibodies

          Invitrogen

          CITED2 Polyclonal Antibody

          View all (11) CITED2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CITED2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CITED2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CITED2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CITED2 Antibody (PA5-114719) in ICC/IF

          Immunocytochemistry analysis of CITED2 in HeLa cells. Samples were treated with PFA, permeabilized in 0.1% Triton X-100, blocked in 10% serum (45 min at 25°C), and incubated with polyclonal antibody (Product # PA5-114719). Secondary staining was applied with mouse anti-beta tubulin (1 hr at 37°, AlexaFluor 594 conjugated goat anti-rabbit IgG (Red), AlexaFluor 488 conjugated goat anti-mouse IgG (Green) and DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CITED2 Antibody in Immunocytochemistry (ICC/IF)
          CITED2 Antibody in Western Blot (WB)
          CITED2 Polyclonal Antibody

          Product Details

          PA5-114719

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CITED2(Accession Q99967), corresponding to amino acid residues Y134-G184.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2899355

          Product Specific Information

          Antibody detects endogenous levels of total CITED2.

          Target Information

          The protein encoded by this gene inhibits transactivation of HIF1A-induced genes by competing with binding of HIF1a to p300-CH1. Mutations in this gene are a cause of cardiac septal defects. Three transcript variants encoding the same protein have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cbp/p300-interacting transactivator 2; melanocyte specific gene 2; melanocyte-specific gene 1-related gene 1; MRG-1; MSG-related protein 1; MSG1-related gene 1; P35srj

          View more View less

          Gene Aliases: AI835299; ASD8; CITED2; ER154-like; MRG-1; MRG1; Msg2; P35SRJ; VSD2

          View more View less

          UniProt ID: (Human) Q99967, (Mouse) O35740

          View more View less

          Entrez Gene ID: (Human) 10370, (Rat) 114490, (Mouse) 17684

          View more View less

          Function(s)
          chromatin binding transcription coactivator activity transcription corepressor activity protein binding protein domain specific binding histone acetyltransferase binding SMAD binding LBD domain binding RNA polymerase II sequence-specific DNA binding transcription factor binding molecular function activator activity RNA polymerase II activating transcription factor binding RNA polymerase II transcription coactivator activity RNA polymerase II transcription corepressor activity molecular_function transcription factor activity, sequence-specific DNA binding transcription cofactor activity transcription cofactor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter blood vessel development vasculogenesis response to hypoxia in utero embryonic development trophectodermal cell differentiation neural tube formation neural tube closure liver development embryonic placenta development vasculature development heart looping lens morphogenesis in camera-type eye hematopoietic progenitor cell differentiation leukocyte differentiation determination of left/right asymmetry in lateral mesoderm outflow tract morphogenesis regulation of organ formation endocardial cushion development ventricular septum development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transforming growth factor beta receptor signaling pathway determination of left/right symmetry central nervous system development peripheral nervous system development heart development sex determination cell proliferation male gonad development response to mechanical stimulus positive regulation of gene expression negative regulation of gene expression cranial nerve morphogenesis positive regulation of cell-cell adhesion cell differentiation adrenal gland development negative regulation of cell migration positive regulation of transforming growth factor beta receptor signaling pathway granulocyte differentiation response to fluid shear stress positive regulation of peroxisome proliferator activated receptor signaling pathway adrenal cortex formation skeletal muscle cell differentiation nodal signaling pathway negative regulation of apoptotic process response to estrogen positive regulation of cell cycle negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter decidualization spleen development thymus development embryonic camera-type eye morphogenesis erythrocyte development uterus development embryonic process involved in female pregnancy bone morphogenesis cardiac septum morphogenesis ventricular septum morphogenesis embryonic heart tube left/right pattern formation left/right pattern formation pulmonary artery morphogenesis cardiac neural crest cell development involved in heart development determination of heart left/right asymmetry left/right axis specification cellular response to growth factor stimulus cellular response to hypoxia cellular senescence positive regulation of male gonad development branching involved in ureteric bud morphogenesis kidney development morphogenesis of an epithelium cell aging negative regulation of transcription from RNA polymerase II promoter in response to hypoxia nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry regulation of nucleic acid-templated transcription transcription, DNA-templated transcription from RNA polymerase II promoter multicellular organism development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline