Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Antibody detects endogenous levels of total CAP1.
Adenylyl cyclase-associated protein 1 is an enzyme that in humans is encoded by the CAP1 gene. This gene is mapped to 1p34. 2. The protein encoded by this gene is related to the S. cerevisiae CAP protein, which is involved in the cyclic AMP pathway. And the human protein is able to interact with other molecules of the same protein, as well as with CAP2 and actin. Alternatively spliced transcript variants have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; Adenylyl cyclase-associated protein 1; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP1; cyclase-associated protein homologue; RP11-115D7.1; testis secretory sperm-binding protein Li 218p; unnamed protein product
Gene Aliases: CAP; CAP1; CAP1-PEN; Mch1
UniProt ID: (Human) Q01518, (Rat) Q08163, (Mouse) P40124
Entrez Gene ID: (Human) 10487, (Rat) 64185, (Mouse) 12331
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support