Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 14
Antibody detects endogenous levels of total ILF1.
The protein encoded by this gene contains a fork head DNA binding domain. This protein can bind to the purine-rich motifs of the HIV long terminal repeat (LTR), and to the similar purine-rich motif in the interleukin 2 (IL2) promoter. It may be involved in the regulation of viral and cellular promoter elements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cellular transcription factor ILF-1; Forkhead box protein K2; FOXK1; foxk2; G/T-mismatch specific binding protein; interleukin enhancer binding factor 1; Interleukin enhancer-binding factor 1; nGTBP
Gene Aliases: 1110054H05Rik; 5730434B08Rik; 6230415M23Rik; FOXK2; ILF; ILF-1; ILF1; nGTBP
UniProt ID: (Human) Q01167, (Mouse) Q3UCQ1
Entrez Gene ID: (Human) 3607, (Rat) 303753, (Mouse) 68837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support