Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Antibody detects endogenous levels of total KDM6A / UTX.
This gene is located on the X chromosome and is the corresponding locus to a Y-linked gene which encodes a tetratricopeptide repeat (TPR) protein. The encoded protein of this gene contains a JmjC-domain and catalyzes the demethylation of tri/dimethylated histone H3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4lysine (K)-specific demethylase 6A; [histone H3]-trimethyl-L-lysine(27) demethylase 6A; bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeat protein (UTX)); Histone demethylase UTX; lysine (K)-specific demethylase 6A; Lysine-specific demethylase 6A; ubiquitously transcribed tetratricopeptide repeat gene, X chromosome; ubiquitously transcribed tetratricopeptide repeat protein X-linked; Ubiquitously transcribed TPR protein on the X chromosome; Ubiquitously transcribed X chromosome tetratricopeptide repeat protein; ubiquitously-transcribed TPR gene on the X chromosome; Ubiquitously-transcribed TPR protein on the X chromosome; Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein; unnamed protein product; v3; v4; v5; v6; v7
Gene Aliases: bA386N14.2; KABUK2; KDM6A; UTX
UniProt ID: (Human) O15550, (Mouse) O70546
Entrez Gene ID: (Human) 7403, (Mouse) 22289, (Rat) 103689969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support