Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Antibody detects endogenous levels of total ZW10.
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centromere/kinetochore protein zw10 homolog; H KNTC1AP; MGC149821; unnamed protein product; zeste white 10 homolog; ZW10 homolog, centromere/kinetochore protein; ZW10, kinetochore associated, homolog
Gene Aliases: 6330566F14Rik; HZW10; KNTC1AP; MmZw10; ZW10
UniProt ID: (Human) O43264, (Mouse) O54692, (Rat) Q4V8C2
Entrez Gene ID: (Human) 9183, (Mouse) 26951, (Rat) 363059
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support