Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HMSPase; Microsomal signal peptidase 21 kDa subunit; SEC11 homolog C; SEC11-like 3; SEC11-like protein 3; signal peptidase complex 21; Signal peptidase complex catalytic subunit SEC11C; SPase 21 kDa subunit; SPC21; unnamed protein product
Gene Aliases: 1810029G24Rik; SEC11C; SEC11L3; SPC21; SPCS4C
UniProt ID: (Human) Q9BY50, (Mouse) Q9D8V7
Entrez Gene ID: (Human) 90701, (Mouse) 66286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support