Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Positive Samples: Mouse heart, Mouse liver, Rat liver
Immunogen sequence: MASILRSPQA LQLTLALIKP DAVAHPLILE AVHQQILSNK FLIVRMRELL WRKEDCQRFY REHEGRFFYQ RLVEFMASGP IRAYILAHKD AIQLWRTLMG PTRVFRARHV APDSIRGSFG LTDTRNTTHG SDSVVSASRE IAAFFPDFSE QRWYEEEEPQ LRCGPVCYSP EGGVHYVAGT GGLGPA
Nucleoside diphosphate (NDP) kinases (EC 2.7.4.6), such as NME6, are ubiquitous enzymes that catalyze transfer of gamma-phosphates, via a phosphohistidine intermediate, between nucleoside and dioxynucleoside tri- and diphosphates.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: expressed in non-metastatic cells 6 (nucleoside-diphosphate kinase); expressed in non-metastatic cells 6 protein (nucleoside diphosphate kinase); expressed in non-metastatic cells 6, protein (nucleoside diphosphate kinase); Inhibitor of p53-induced apoptosis-alpha; IPIA-alpha; NDK 6; NDK6; NDP kinase 6; nm23-H6; nm23-M6; nm23-R6; nm23/nucleoside diphosphate kinase 6; non-metastatic cells 6, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase 6; Nucleoside diphosphate kinase 6, mitochondrial; nucleoside diphosphate kinase type 6; unnamed protein product
Gene Aliases: IPIA-ALPHA; NDK 6; NM23-H6; nm23-M6; Nm23-R6; NME6
UniProt ID: (Human) O75414, (Mouse) O88425, (Rat) O88426
Entrez Gene ID: (Human) 10201, (Mouse) 54369, (Rat) 58964
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support