Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: BT-474, HT-29, Mouse pancreas
Immunogen sequence: MMGSLLAATT EAPGEYFFSD GVRLKKYRGM GSLDAMEKSS SSQKRYFSEG DKVKIAQGVS GSIQDKGSIQ KFVPYLIAGI QHGCQDIGAR SLSVLRSMMY SGELKFEKRT MSAQIEGGVH GLHSYEKRLY
The protein encoded by this gene acts as a homotetramer to regulate cell growth. The encoded protein is an enzyme that catalyzes the synthesis of xanthine monophosphate (XMP) from inosine-5'-monophosphate (IMP). This is the rate-limiting step in the de novo synthesis of guanine nucleotides. Defects in this gene are a cause of retinitis pigmentosa type 10 (RP10). Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781N0678; IMP (inosine 5'-monophosphate) dehydrogenase 1; IMP (inosine monophosphate) dehydrogenase 1; IMP dehydrogenase 1; IMP dehydrogenase 1 {ECO:0000255|HAMAP-Rule:MF_03156}; IMP dehydrogenase type 1 (EC 1.1.1.205); IMPD 1; IMPD 1 {ECO:0000255|HAMAP-Rule:MF_03156}; IMPDH 1; IMPDH 1 {ECO:0000255|HAMAP-Rule:MF_03156}; IMPDH-I; inosine 5'-phosphate dehydrogenase 1; inosine monophosphate dehydrogenase 1; Inosine-5'-monophosphate dehydrogenase 1; inosine-5'-monophosphate dehydrogenase 1 {ECO:0000255|HAMAP-Rule:MF_03156}; unnamed protein product
Gene Aliases: B930086D20Rik; IMPD; IMPD 1; IMPD1; IMPDH 1; IMPDH-I; IMPDH1; LCA11; RP10; sWSS2608
UniProt ID: (Human) P20839, (Rat) D3ZLZ7, (Mouse) P50096
Entrez Gene ID: (Human) 3614, (Rat) 362329, (Mouse) 23917
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support