Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: HT-29, HeLa, Mouse lung, Mouse thymus, Mouse spleen, Rat lung, Rat thymus
Immunogen sequence: MAEIIQERIE DRLPELEQLE RIGLFSHAEI KAIIKKASDL EYKIQRRTLF KEDFINYVQY EINLLELIQR RRTRIGYSFK KDEIENSIVH RVQGVFQRAS AKWKDDVQLW LSYVAFCKKW ATKTRLSKVF SAMLAIHSNK PALWIMAAKW EMEDRLSSES ARQLFLRALR FHPECPKLYK EYFRMELMHA EKLRKEKEEF EKASMDVENP DYSEEILKGE
Involved in nucleolar processing of pre-18S ribosomal RNA. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma-associated antigen 66; Multiple hat domains protein; predicted protein of HQ1289; U3 small nucleolar RNA-associated protein 6 homolog; unnamed protein product; UTP6, small subunit (SSU) processome component, homolog; UTP6, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases: 4732497O03Rik; C17orf40; HCA66; MHAT; RGD1311017; UTP6
UniProt ID: (Human) Q9NYH9, (Mouse) Q8VCY6
Entrez Gene ID: (Human) 55813, (Rat) 360574, (Mouse) 216987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support