Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Samples: U-87MG, Mouse ovary, Mouse lung
Immunogen sequence: CNSSSALPLY NQQVGDCCII RVSLDVDNGN MYKSILVTSQ DKAPAVIRKA MDKHNLEEEE PEDYELLQIL SDDRKLKIPE NANVFYAMNS TANYDFVLKK RTFTKGVKVK HGASSTLPRM KQKGLKIAKG IF
Stimulates the dissociation of GDP from the Ras-related RalA and RalB GTPases which allows GTP binding and activation of the GTPases. Interacts and acts as an effector molecule for R-Ras, H-Ras, K-Ras, and Rap.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ral guanine nucleotide dissociation stimulator; Ral guanine nucleotide exchange factor; RalGDS; RalGEF; unnamed protein product
Gene Aliases: Gnds; KIAA1308; mKIAA1308; RALGDS; RalGEF; RGDS; RGF
UniProt ID: (Human) Q12967, (Mouse) Q03385, (Rat) Q03386
Entrez Gene ID: (Human) 5900, (Mouse) 19730, (Rat) 29622
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support