Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Samples: U-87MG, A-549, A-431, Rat lung, Rat heart
Immunogen sequence: WDYRMNQKLW EVYSGHPVQH ISFSSHSLIT ANVPYQTVMR NADLDSFTTH RRHRGLIRAY EFAVDQLAFQ SPLPVCRSSC DAMATHYYDL ALAFPYNHV
This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains a WD-40 domain, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: F-box and WD-40 domain protein 8; F-box and WD-40 domain-containing protein 8; F-box only protein 29; F-box protein FBX29; F-box/WD repeat-containing protein 8; MGC33534; unnamed protein product
Gene Aliases: 4930438M06Rik; FBW6; FBW8; FBX29; FBXO29; FBXW6; FBXW8
UniProt ID: (Human) Q8N3Y1, (Mouse) Q8BIA4
Entrez Gene ID: (Human) 26259, (Rat) 304522, (Mouse) 231672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support