Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: SKOV3, 293T, Mouse testis, Mouse pancreas, Rat large intestine, Rat kidney
Immunogen sequence: HMQESPHKCP TCGRTFNQRS NLKTHLLTHT DIKPYSCEQC GKVFRRNCDL RRHSLTHTPR QDF
OSR2 is a mammalian homolog of the Drosophila odd-skipped family of transcription factors.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: odd-skipped related 2; OSR2A; zing finger protein; OSR2B; zinc finger protein; Protein odd-skipped-related 2; unnamed protein product
Gene Aliases: 5430409I15Rik; OSR2; Osr2A; Osr2B
UniProt ID: (Human) Q8N2R0, (Mouse) Q91ZD1, (Rat) Q6AY34
Entrez Gene ID: (Human) 116039, (Mouse) 107587, (Rat) 315039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support