Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive Samples: U-251MG, Mouse brain
Immunogen sequence: LQGLQVLLPD YLQERFVQAA LSYIACNSEG EFICKENDCW CHCGPKFPEC NCPSMDIQAM EENLLRITET WKAYNSDFEE SDEFKLFMKR LPMNYFLNTS TIMHLWTMDS NFQRRYEQLE NSMKQLFLKA QKIVHKLFSL SKRCHKQPLI SLPRQRTSTY WLTRIQSFLY CNENGLLGSF SEETHSCTCP NDQVVCTAFL PCTVGDASAC LTCAPDNRTR CGTCNTGYML SQGLCKPEVA ESTDHYIGFE TDLQDLEMKY L
Inhibits neuronal cell proliferation by negative regulation of the cell cycle transition. Promotes pituitary gonadotrope cell proliferation, migration and invasion, when overexpressed. May play a role in cell pituitary tumor development.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP/retinoic acid-inducible neural-specific protein 3; bone morphogenetic protein/retinoic acid inducible neural-specific 3; DBCCR1-like protein 1; family with sequence similarity 5, member C; protein FAM5C; RP11-445K1.1; unnamed protein product
Gene Aliases: B830045N13Rik; BRINP3; DBCCR1L; DBCCR1L1; FAM5C
UniProt ID: (Human) Q76B58, (Rat) Q8K1M7, (Mouse) Q499E0
Entrez Gene ID: (Human) 339479, (Rat) 286901, (Mouse) 215378
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support