Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: HeLa, HepG2, K-562, NIH/3T3, C6, Mouse liver, Mouse lung, Rat brain, Rat testis
Immunogen sequence: MAAADGDDSL YPIAVLIDEL RNEDVQLRLN SIKKLSTIAL ALGVERTRSE LLPFLTDTIY DEDEVLLALA EQLGTFTTLV GGPEYVHCLL PPLESLATVE ETVVRDKAVE SLRAISHEHS PSDLEAHFVP LVKRLAGGDW FTSRTSACGL FSVCYPRVSS A
Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit.The B regulatory subunit might modulate substrate selectivity and catalytic activity.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha isoform of regulatory subunit A, protein phosphatase 2; B55; Medium tumor antigen-associated 61 kDa protein; PP2A subunit A isoform PR65-alpha; PP2A subunit A isoform PR65-beta; PP2A subunit A isoform R1-alpha; PP2A subunit A isoform R1-beta; PP2A subunit B isoform alpha; PP2A subunit B isoform B55-alpha; PP2A subunit B isoform BRA; PP2A subunit B isoform PR55-alpha; PP2A subunit B isoform R2-alpha; PP2A, subunit A, PR65-beta isoform; PP2A, subunit A, R1-beta isoform; PP2A, subunit B, B-alpha isoform; PP2A, subunit B, B55-alpha isoform; PP2A, subunit B, BRA isoform; PP2A, subunit B, PR55-alpha isoform; PP2A, subunit B, R2-alpha isoform; PP2a-Abeta; protein phosphatase PP2A regulatory/structural subunit A isoform beta; PP2Aa; PPP2R1A; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alpha isoform; protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform; protein phosphatase 2, regulatory subunit A, alpha; protein phosphatase 2, regulatory subunit A, beta; protein phosphatase 2, regulatory subunit B, alpha; protein phosphatase 2, structural/regulatory subunit A, beta; protein phosphatase PP2A; serine/threonine protein phosphatase A subunit type 2A; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alpha isoform; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A beta isoform; testicular secretory protein Li 1; unnamed protein product
Gene Aliases: 2410004D02Rik; 2410091N08Rik; 6330556D22Rik; AI790395; B55; B55A; B55ALPHA; HJS2; mKIAA1541; MRD36; PP2A; PP2A-Aalpha; PP2A-Abeta; PP2AA; PP2AAALPHA; PPP2R1A; PPP2R1B; PPP2R2A; PR52A; PR55A; PR55alpha; PR65; PR65A; PR65B
UniProt ID: (Human) P30153, (Human) P30154, (Human) P63151, (Mouse) Q76MZ3, (Mouse) Q7TNP2, (Rat) Q4QQT4, (Mouse) Q6P1F6, (Rat) P36876
Entrez Gene ID: (Human) 5518, (Human) 5519, (Human) 5520, (Mouse) 51792, (Rat) 117281, (Mouse) 73699, (Rat) 315648, (Mouse) 71978, (Rat) 117104
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support