Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Positive Samples: A375, A431, HepG2, TE-1, SGC7901, mouse stomach, mouse liver, rat liver
Immunogen sequence: MSTLLENIFA IINLFKQYSK KDKNTDTLSK KELKELLEKE FRQILKNPDD PDMVDVFMDH LDIDHNKKID FTEFLLMVFK LAQAYYESTR KE
Filaggrin is expressed only in well differentiated keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epidermal filaggrin; Filaggrin; Filaggrin (profilaggrin)
Gene Aliases: ATOD2; AW107830; FLG; FLG-1; FLG1; ft; Pflg
UniProt ID: (Human) P20930
Entrez Gene ID: (Human) 2312, (Rat) 24641, (Mouse) 14246
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support