Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive Samples: Mouse brain
Immunogen sequence: GVAAPPVPPP PPTPFPQAKA RRKGRRGGKC STRCFCPRPH AKAFACPVES CVRSFARSDE LNRHLRIHTG HKPFQCRICL RNFSRSDHLT THVRTHTGEK PFACDVCGRR FARSDEKKRH SKVHLKQKAR AEERLKGLGF YSLGLSFASL
Early growth response protein 4 (EGR-4), also known as AT133, is a protein that in humans is encoded by the EGR4 gene. EGR-4 is a member of the early growth response (EGF) family of zinc finger transcription factors. Transcriptional regulator. Recognizes and binds to the DNA sequence 5'-GCGGGGGCG-3' (GSG). Activates the transcription of target genes whose products are required for mitogenesis and differentiation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AT133; Early growth response protein 4; Early response protein NGFI-C; EGR-4; Nerve growth factor-induced protein C; NGFI-C; unnamed protein product; zinc-finger transcription factor NGFI-C (early response gene)
Gene Aliases: AT133; Egr-4; EGR4; Egr4l1; NGF1-C; NGFI-C; NGFIC; PAT133
UniProt ID: (Human) Q05215, (Rat) Q00911, (Mouse) Q9WUF2
Entrez Gene ID: (Human) 1961, (Rat) 25129, (Mouse) 13656
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support