Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody has been tested in direct-ELISA
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin-like 7A; actin-like 7-alpha; Actin-like protein 9; testicular tissue protein Li 15; testicular tissue protein Li 9; unnamed protein product
Gene Aliases: 1700029I08Rik; ACTL7C; ACTL9; AV259599; HSD21; SPGF53
UniProt ID: (Human) Q8TC94, (Mouse) Q8CG27
Entrez Gene ID: (Human) 284382, (Mouse) 69481
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support