Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: HepG2, Mouse brain, Rat brain. The target is usually found in the following locations: Secreted.
Immunogen sequence: GQRWELALGR FWDYLRWVQT LSEQVQEELL SSQVTQELRA LMDETMKELK AYKSELEEQL TPVAEETRAR LSKELQAAQA RLGADMEDVC GRLVQYRGEV QAMLGQSTEE LRVRLASHLR KLRKRLLRDA DDLQKRLAVY QAGAREGAER GLSAIRERLG PLVEQGRVRA ATVGSLAGQP LQERAQAWGE RLRARMEEMG SRTRDRLDEV KEQVAEVRAK LEEQAQQIRL QAEAFQARLK SWFEPLVEDM QRQWAGLVEK VQAAVGT
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13. 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-E; ApoE4; Apolipoprotein; Apolipoprotein E; apolipoprotein E3; apolipoprotein E4; Apoprotein; B2G1; BG; MGC1571; unnamed protein product
Gene Aliases: AD2; APO-E; APOE; ApoE4; APOEA; LDLCQ5; LPG
UniProt ID: (Human) P02649, (Rat) P02650
Entrez Gene ID: (Human) 348, (Rat) 25728
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support