Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: 293T, U-251MG, HeLa, COS-7, PC-12, Jurkat, Mouse liver, Mouse kidney. The target is usually found in the following locations: Cytoplasm, Nucleus, P-body.
Immunogen sequence: MSRRYDSRTT IFSPEGRLYQ VEYAMEAIGH AGTCLGILAN DGVLLAAERR NIHKLLDEVF FSEKIYKLNE DMACSVAGIT SDANVLTNEL RLIAQRYLLQ YQEPIPCEQL VTALCDIKQA YTQFGGKRPF GVSLLYIGWD KHYGFQLYQS DPSGNYGGWK ATCIGNNSAA AVSMLKQDYK EGEMTLKSAL ALAIKVLNKT MDVSKLSAEK VEIATLTREN GKTVIRVLKQ KEVEQLIKKH EEEEAKAERE KKEKEQKEKD K
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-3; Macropain subunit C9; MGC111191; MGC12467; MGC24813; Multicatalytic endopeptidase complex subunit C9; multicatalytic proteinase subunit L; proteasome (prosome, macropain) subunit, alpha type, 4; proteasome alpha 4 subunit; Proteasome component C9; proteasome subunit alpha 4; Proteasome subunit alpha type-4; Proteasome subunit alpha-3; proteasome subunit alpha3; proteasome subunit HC9; Proteasome subunit L; PSMA 4; unnamed protein product
Gene Aliases: C9; HC9; HsT17706; PSC9; PSMA4
UniProt ID: (Human) P25789, (Mouse) Q9R1P0, (Rat) P21670
Entrez Gene ID: (Human) 5685, (Mouse) 26441, (Rat) 29671
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support