Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • SRD5A1 Antibodies

          Invitrogen

          SRD5A1 Polyclonal Antibody

          View all (8) SRD5A1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SRD5A1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SRD5A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SRD5A1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SRD5A1 Antibody (PA5-120297) in WB

          Western blot analysis of SRD5A1 in extracts of various cell lines. Samples were incubated with SRD5A1 Polyclonal antibody (Product # PA5-120297) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Enhanced Kit. Exposure time: 90s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SRD5A1 Antibody in Western Blot (WB)
          SRD5A1 Polyclonal Antibody

          Product Details

          PA5-120297

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.57 mg/mL

          Amount

          157 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2913869

          Product Specific Information

          Positive test controls include: Mouse brain, Mouse liver, Mouse testis, Rat liver. The target is usually found in the following locations: Endoplasmic reticulum membrane, Microsome membrane, Multi-pass membrane protein.

          Immunogen sequence: LIYPFLMRGG KPMPLLACTM AIMFCTCNGY LQSRYLSHCA VYADDWVTDP RFLIGFGLWL TGMLINIHSD HILRNLRKPG DTGYKIPRGG LFEYVTAANY F

          Target Information

          Steroid 5alpha-Reductase is an important enzyme in androgen physiology because it catalyzes the conversion of testosterone into the more potent 5alpha-dihydro-testosterone, which mediates androgen effects on target tissues. The enzyme exists as two isoforms: type 1, which is expressed mainly in the skin; and type 2, which is expressed mainly in the prostate. In cultured human skin cells, 5alpha-Reductase 1 shows heterogeneity of protein, and has different levels of transcriptional and translational expression. 5alpha-Reductase 1 is expressed in all portions of the hair follicle, whereas 5alpha-Reductase 2 is expressed only in mesenchymal portions. In addition, 5alpha-Reductase 1 is mainly expressed in human breast carcinoma and may play a role in the in situ production and actions of the potent androgen 5alpha-dihydrotestosterone, including inhibition of cancer cell proliferation in hormone-dependent human breast carcinoma. The 5alpha-Reductase-3alpha-hydroxysteroid dehydrogenase complex is present in the human brain, suggesting that the complex may be involved in the synthesis of neuroactive steroids or the catabolism of neurotoxic steroids.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 3-oxo-5-alpha-steroid 4-dehydrogenase 1; S5AR 1; SR type 1; steroid 5 alpha-reductase 1; Steroid 5-alpha-reductase 1; steroid-5-alpha-reductase 1; type 1 5alpha-reductase

          View more View less

          Gene Aliases: 0610031P22Rik; 4930435F02Rik; S5AR 1; Srd5a-1; Srd5a1

          View more View less

          UniProt ID: (Mouse) Q68FF9, (Rat) P24008

          View more View less

          Entrez Gene ID: (Mouse) 78925, (Rat) 24950

          View more View less

          Function(s)
          3-oxo-5-alpha-steroid 4-dehydrogenase activity oxidoreductase activity oxidoreductase activity, acting on the CH-CH group of donors amide binding cholestenone 5-alpha-reductase activity NADPH binding dehydrogenase
          Process(es)
          lipid metabolic process steroid biosynthetic process androgen biosynthetic process sex differentiation steroid metabolic process androgen metabolic process cell differentiation progesterone metabolic process response to drug oxidation-reduction process urogenital system development liver development male gonad development cellular response to starvation response to organic cyclic compound response to muscle activity diterpenoid metabolic process spinal cord development hippocampus development thalamus development hypothalamus development pituitary gland development cerebral cortex development male genitalia development female genitalia development response to follicle-stimulating hormone response to estradiol cellular response to insulin stimulus response to testosterone serotonin metabolic process circadian sleep/wake cycle, REM sleep response to estrogen male sex differentiation bone development response to growth hormone response to fungicide cellular response to cAMP cellular response to growth factor stimulus cellular response to estradiol stimulus cellular response to testosterone stimulus cellular response to organic cyclic compound cellular response to dexamethasone stimulus cellular response to epinephrine stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline