Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: K-562. The target is usually found in the following locations: Cell membrane, Single-pass type II membrane protein.
Immunogen sequence: VTLKIEMKKM NKLQNISEEL QRNISLQLMS NMNISNKIRN LSTTLQTIAT KLCRELYSKE QEHKCKPCPR RWIWHKDSCY FLSDDVQTWQ ESKMACAAQN ASLLKINNKN ALEFIKSQSR SYDYWLGLSP EEDSTRGMRV DNIINSSAWV IRNAPDLNNM YCGYINRLYV QYYHCTYKKR MICEKMANPV QLGSTYFREA
CD371 is also known as Clec12A or Myeloid Inhibitory C-type Lectin-like receptor (MICL). CD371 is a highly glycosylated transmembrane receptor with a cytoplasmic ITIM that associates with SHP1 and SHP2. CD371 is expressed mainly on granulocytes, monocytes, macrophages, and dendritic cells, including plasmacytoid dendritic cells. Expression of CD371 on NK cells remains controversial. Although the function of CD371 is not fully understood, studies suggest that this receptor plays a role in inflammation and LPS activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 12 member A; C-type lectin protein CLL-1; C-type lectin superfamily; C-type lectin-like molecule 1; C-type lectin-like molecule-1; CD371; CLL-1; DCAL-2; Dendritic cell-associated lectin 2; hKLRL1; Killer cell C-type lectin-like receptor L1; MICL; MICL-alpha; type II transmembrane protein; MICL-beta; type II transmembrane protein; MICL-gamma; type II transmembrane protein; mKLRL1; Myeloid inhibitory C-type lectin-like receptor; unnamed protein product
Gene Aliases: CD371; CLEC12A; CLL-1; CLL1; D230024O04; DCAL-2; DCAL2; hKLRL1; KLRL1; MICL
UniProt ID: (Human) Q5QGZ9, (Mouse) Q504P2
Entrez Gene ID: (Human) 160364, (Mouse) 232413, (Rat) 680338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support