Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: SKOV3, MCF7, HepG2, Mouse kidney, Mouse lung, Mouse liver, Mouse thymus, Rat lung, Rat kidney. The target is usually found in the following locations: Endoplasmic reticulum membrane, Single-pass membrane protein.
Immunogen sequence: RLLFSKTYKL QERSDLTVKE KEELIEEWQP EPLVPPVPKD HPALNYNIVS GPPSHKTVVN GKECINFASF NFLGLLDNPR VKAAALASLK KYGVGTCGPR GFYGTFDVHL DLEDRLAKFM KTEEAIIYSY GFATIASAIP AYSKRGDIVF VDRAACFAIQ KGLQASRSDI KLFKHNDMAD LERLLKEQEI EDQKNPRKAR VTRRFIVVEG LYMNTGTICP LPELVKLKYK YKARIFLEES LSFGVLGEHG RGVTEHYGIN IDDIDLISAN MENA
Serine palmitoyltransferase (SPT) is the key enzyme in sphingolipid biosynthesis. It catalyzes the pyridoxal-5-prime-phosphate-dependent condensation of L-serine and palmitoyl-CoA to 3-oxosphinganine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hLCB1; HSAN; LCB 1; Long chain base biosynthesis protein 1; MGC14645; RP11-118F2.1; serine C-palmitoyltransferase; Serine palmitoyltransferase 1; serine palmitoyltransferase subunit 1; Serine-palmitoyl-CoA transferase 1; SPT 1; SPT1; unnamed protein product
Gene Aliases: ALS27; AW552086; C77762; E030036H05; HSAN1; HSN1; LBC1; LCB1; rCG_44191; RGD1306617; SPT1; SPTI; SPTLC1
UniProt ID: (Human) O15269, (Mouse) O35704, (Rat) D4A2H2
Entrez Gene ID: (Human) 10558, (Mouse) 268656, (Rat) 361213
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support