Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: Rat lung, Rat kidney. The target is usually found in the following locations: Nucleus.
Immunogen sequence: EEYYPAFEHQ TACDSYWTSV HPEYWTKRHV WEWLQFCCDQ YKLDTNCISF CNFNISGLQL CSMTQEEFVE AAGLCGEYLY FILQNIRTQG YSFFNDAEES KATIKDYADS N
ELF5 is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E74-like factor 5; E74-like factor 5 (ets domain transcription factor); elf-5; Epithelium specific Ets-2; Epithelium-restricted ESE-1-related Ets factor; Epithelium-specific Ets transcription factor 2; ESE-2; Ets transcription factor; ETS-related transcription factor Elf-5; unnamed protein product
Gene Aliases: ELF5; ESE-2; ESE-5; ESE-5.; ESE2
UniProt ID: (Human) Q9UKW6, (Mouse) Q8VDK3
Entrez Gene ID: (Human) 2001, (Mouse) 13711, (Rat) 366142
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support