Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: A-549, HT-29, Mouse liver, Mouse testis, Mouse lung. The target is usually found in the following locations: Nucleus.
Immunogen sequence: MSKISQQNST PGVNGISVIH TQAHASGLQQ VPQLVPAGPG GGGKAVAPSK QSKKSSPMDR NSDEYRQRRE RNNMAVKKSR LKSKQKAQDT LQRVNQLKEE NERLEAKIKL LTKELSVLKD LFLEHAHNLA DNVQSISTEN TTADGDNAGQ
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C EBP; C/EBP gamma; CCAAT/enhancer binding protein (C/EBP), gamma; CCAAT/enhancer-binding protein gamma; CEBP gamma; G-CSF promoter element 1 binding protein; GPE1-binding protein; GPE1-BP; GPE1-BP (C/EBP gamma); Granulocyte colony-stimulating factor promoter element 1-binding protein; IG/EBP-1; Immunoglobulin enhancer-binding protein 1; unnamed protein product
Gene Aliases: C/EBP[g]; C/EBPgamma; CEBPG; GPE1-BP; GPE1BP; Ig/EBP; IG/EBP-1
UniProt ID: (Human) P53567, (Mouse) P53568
Entrez Gene ID: (Human) 1054, (Mouse) 12611
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support