Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: HeLa, Mouse kidney, Mouse liver, Mouse brain, Rat brain. The target is usually found in the following locations: Cell junction, Cell projection, Cytoplasm, Golgi apparatus, Golgi apparatus membrane, Peripheral membrane protein, dendrite, postsynaptic cell membrane, postsynaptic density, synapse, trans-Golgi network membrane.
Immunogen sequence: ARLAAKYLDK ELAGRVQQIQ LLGRDMKGPA HDKLWNQLEA EIHLHRHKTV IRACRGRNDL KRPMQAPPGH DQDSLKKSQG VGPIRKVLLL KEDHEGLGIS ITGGKEHGVP ILISEIHPGQ PADRCGGLHV GDAILAVNGV NLRDTKHKEA VTILSQQRGE IEFEVVYVAP EVDSDDENVE YEDESGHRYR LYLDELEGGG NPGASCKDTS GEIKVLQGFN KKAVTDTHEN GDLGTASETP LDDGASKLDD LHTLYHKKSY
PDZ domains contain approximately 90 amino acids and bind the extreme C terminus of proteins in a sequence-specific manner. PIST, a PDZ domain-containing Golgi protein, was discovered in a yeast two-hybrid system as a binding partner to Beclin-1, a Bcl-2-interacting protein homologous to the yeast autophagy gene apg6. Experiments with mutant PIST proteins lacking the PDZ domain showed that PIST interaction with Beclin-1 through its coiled-coil domain can modulate Beclin-1 activity and suggest that PIST interactions with other proteins through its PDZ domain may regulate the activity of PIST and Beclin-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFTR-associated ligand; dJ94G16.2 PIST; FIG; PDZ domain-containing Golgi protein; Fused in glioblastoma; golgi-associated PDZ and coiled-coil motif containing protein; Golgi-associated PDZ and coiled-coil motif-containing protein; PDZ domain-containing protein; PDZ domain-containing protein PIST; PDZ protein interacting specifically with TC10; PDZ/coiled-coil domain binding partner for the rho-family GTPase TC10; PIST
Gene Aliases: 2210402P09Rik; AI844555; CAL; dJ94G16.2; FIG; GOPC; GOPC1; PIST
UniProt ID: (Human) Q9HD26, (Mouse) Q8BH60
Entrez Gene ID: (Human) 57120, (Mouse) 94221, (Rat) 309774
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support