Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: Mouse intestine, Mouse testis, Mouse liver, Rat lung. The target is usually found in the following locations: Nucleus.
Immunogen sequence: GTPGETVRHC SAPEDPIFRF SSLHSYPFPG TIKSRDMSWK RHHLIPETFG VKRRRKRGPV ESDPLRGEPG SARAAVSELM QLFPRGLFED ALPPIVLRSQ VYSLVPDRTV ADRQLKELQE QGEIRIVQLG FDLDAHGIIF TEDYRTRVLK ACDGRPYAGA VQKFLASVLP ACGDLSFQQD QMTQTFGFRD SEITHLVNAG VLTVRDAGSW WLAVPGAGRF IKYFVKGRQA VLSMVRKAKY RELLLSELLG RRAPVVVRLG LTYHVHDLIG AQLVDCISTT SGTLLRLPET
This gene encodes a serine/threonine kinase which localizes predominantly to the nucleus. Its specific function is unknown; it is possible that phosphorylation of this protein is involved in transcriptional regulation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6 and expresses two transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G11; Inactive serine/threonine-protein kinase 19; MHC class III HLA-RP1; nuclear serine/threonine protein kinase; Protein G11; Protein RP1; serine/threonine kinase 19; serine/threonine-protein kinase 19; Tandem winged helix protein formerly known as STK19; Tandem Winged Helix protein previously known as STK19; unnamed protein product; Winged helix repair factor 1
Gene Aliases: D6S60; D6S60E; G11; HLA-RP1; RP1; STK19; TWH19; WHR1
UniProt ID: (Mouse) Q9JHN8, (Human) P49842
Entrez Gene ID: (Rat) 361800, (Mouse) 54402, (Human) 8859
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support