Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: U-87MG, U-251MG, Jurkat, Mouse kidney, Mouse lung, Mouse spleen, Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, Cytoplasm, cytosol.
Immunogen sequence: KQVVHLNLAE DCMNKFKLNI EKLCKTEQDL ALGTDAEGQK VKDSMRVLLP VLLNKNHDNC DKIRAILLYI FSINGTTEEN LDRLIQNVKI ENESDMIRNW SYLGVPIVPQ SQQGKPLRKD RSAEETFQLS RWTPFIKDIM EDAIDNRLDS KEWPYCSQCP AVWNGSGAVS ARQKPRANYL EDRKNGSKLI VFVIGGITYS EVRCAYEVSQ AHKSCEVIIG STHVLTPKKL LDDIKMLNKP KDKVSLIKDE
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mammalian homolog of Unc-18c; MUNC-18-3; Munc-18c; Platelet Sec1 protein; Protein unc-18 homolog 3; Protein unc-18 homolog C; PSP; syntaxin 4 binding protein; syntaxin binding protein 3A; Syntaxin-binding protein 3; Syntaxin-BP3; unc-18 homolog 3; Unc-18C; Unc18-3; unnamed protein product
Gene Aliases: Munc-18c; MUNC18-3; Munc18-c; MUNC18C; PSP; STXBP3; Stxbp3a; Sxtbp3; UNC-18C; Unc18-3; Unc18c
UniProt ID: (Human) O00186, (Mouse) Q60770
Entrez Gene ID: (Human) 6814, (Rat) 114095, (Mouse) 20912
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support