Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
This antibody is predicted to react with bovine and rat based on sequence homology.
PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dsRNA-binding protein; Interferon-inducible double-stranded RNA-dependent protein kinase activator A; PKR interacting protein; PKR-associated protein X; PKR-associating protein X; PRKRA; Protein activator of the interferon-induced protein kinase; Protein kinase, interferon-inducible double-stranded RNA-dependent activator; unnamed protein product
Gene Aliases: DYT16; HSD-14; HSD14; PACT; PRKRA; RAX
UniProt ID: (Human) O75569
Entrez Gene ID: (Human) 8575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support