Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Endothelin A Receptor Antibodies

          Invitrogen

          Endothelin A Receptor Polyclonal Antibody

          View all (9) Endothelin A Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Endothelin A Receptor Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          Endothelin A Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Endothelin A Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Endothelin A Receptor Antibody (PA5-13432) in ICC/IF

          Immunocytochemistry analysis of Endothelin A Receptor in HepG2 cells. Samples were incubated in Endothelin A Receptor polyclonal antibody (Product # PA5-13432) followed by Alexa Fluor 488-conjugated goat anti-rabbit lgG (green). DAPI was used to stain the cell nuclear (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Endothelin A Receptor Antibody in Immunocytochemistry (ICC/IF)
          Endothelin A Receptor Antibody in Western Blot (WB)
          Endothelin A Receptor Antibody in Flow Cytometry (Flow)
          Endothelin A Receptor Antibody in Flow Cytometry (Flow)
          Endothelin A Receptor Polyclonal Antibody

          Product Details

          PA5-13432

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:50
          -

          Flow Cytometry (Flow)

          1:10-1:50
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide between 125-154 amino acids from the central region of human EDNRA
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          800 µg

          Purification

          Size-exclusion - Dialysis, Ammonium sulfate precipitation

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2277673

          Product Specific Information

          This antibody is predicted to react with mouse and rat based on sequence homology.

          Target Information

          This gene encodes the receptor for endothelin-1, a peptide that plays a role in potent and long-lasting vasoconstriction. This receptor associates with guanine-nucleotide-binding (G) proteins, and this coupling activates a phosphatidylinositol-calcium second messenger system. Polymorphisms in this gene have been linked to migraine headache resistance. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Endothelin A; endothelin receptor subtype A; Endothelin receptor type A; Endothelin-1 receptor; endothelin-1-specific receptor; ET-A; ET-AR; ET-AR delta 3-4; ET-AR delta 4; G protein-coupled receptor; G-protein coupled receptor; unnamed protein product

          View more View less

          Gene Aliases: EDNRA; ET-A; ETA; ETA-R; ETAR; ETRA; hET-AR; MFDA

          View more View less

          UniProt ID: (Human) P25101

          View more View less

          Entrez Gene ID: (Human) 1909

          View more View less

          Function(s)
          phosphatidylinositol phospholipase C activity G-protein coupled receptor activity endothelin receptor activity protein binding
          Process(es)
          mitotic cell cycle angiogenesis patterning of blood vessels response to hypoxia in utero embryonic development blood vessel remodeling response to amphetamine regulation of heart rate heart process glomerular filtration cardiac chamber formation left ventricular cardiac muscle tissue morphogenesis atrial cardiac muscle tissue development cardiac neural crest cell migration involved in outflow tract morphogenesis noradrenergic neuron differentiation cellular calcium ion homeostasis smooth muscle contraction mitochondrion organization signal transduction G-protein coupled receptor signaling pathway activation of adenylate cyclase activity adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration axon guidance heart development respiratory gaseous exchange regulation of blood pressure cell proliferation response to wounding gene expression regulation of glucose transport neural crest cell development neural crest cell differentiation neural crest cell fate commitment artery smooth muscle contraction vascular smooth muscle contraction neuron remodeling heparin metabolic process thyroid gland development neuron projection development glomerulus development cellular response to oxidative stress embryonic heart tube development aorta development vasoconstriction norepinephrine metabolic process middle ear morphogenesis positive regulation of I-kappaB kinase/NF-kappaB signaling cellular response to human chorionic gonadotropin stimulus developmental pigmentation enteric nervous system development sympathetic nervous system development axon extension embryonic skeletal system development neuromuscular process sodium ion homeostasis canonical Wnt signaling pathway head development face development axonogenesis involved in innervation establishment of endothelial barrier pharyngeal arch artery morphogenesis renal sodium ion absorption calcium ion transmembrane transport cellular response to follicle-stimulating hormone stimulus cellular response to luteinizing hormone stimulus protein transmembrane transport glomerular endothelium development glomerular visceral epithelial cell differentiation endothelin receptor signaling pathway endothelin receptor signaling pathway involved in heart process renal albumin absorption vascular smooth muscle cell development mesenchymal cell apoptotic process cellular response to toxic substance sympathetic neuron axon guidance cAMP/PKA signal transduction semaphorin-plexin signaling pathway involved in axon guidance glomerular visceral epithelial cell apoptotic process meiotic cell cycle process involved in oocyte maturation cranial skeletal system development response to acetylcholine regulation of protein localization to cell leading edge
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline