Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.
Immunogen sequence: FVMYLTGTRQ MMSSKHGVRI DV
Highest antigen sequence indentity to the following orthologs: Mouse - 77%, Rat - 77%.
TMEM267 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Transmembrane protein 267; transmembrane protein C5orf28; unnamed protein product
Gene Aliases: C5orf28; TMEM267
UniProt ID: (Human) Q0VDI3
Entrez Gene ID: (Human) 64417
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support