Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
SLC23A3 gene ontology annotations related to this gene include transmembrane transporter activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2-binding protein 3; HPC E2-binding protein 3; Na(+)/L-ascorbic acid transporter 3; Sodium-dependent vitamin C transporter 3; solute carrier family 23 (nucleobase transporters), member 3; Solute carrier family 23 member 3; unnamed protein product
Gene Aliases: E2BP3; SLC23A3; SVCT3; Yspl1
UniProt ID: (Human) Q6PIS1
Entrez Gene ID: (Human) 151295
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support