Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Transcription activator that binds calmodulin in a calcium-dependent manner in vitro (PubMed:12218065). Binds to the DNA consensus sequence 5'-[ACG]CGCG[GTC]-3' (By similarity). Regulates transcriptional activity in response to calcium signals (Probable).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AtCAMTA6; AtSR3; Calmodulin-binding transcription activator 6; CMTA6; EICBP.e; EICBP5; Ethylene-induced calmodulin-binding protein 5; Ethylene-induced calmodulin-binding protein e; Signal-responsive protein 3; SR3
Gene Aliases: AT3G16940; CAMTA6; CMTA6; K14A17.5; K14A17_6; SR3
UniProt ID: (Arabidopsis) Q9LSP8
Entrez Gene ID: (Arabidopsis) 820950
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support