Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Trypsin is a 24 kDa enzyme of the serine proteinase family. It is produced in the pancreas as an inactive precursor, trypsinogen, but the active enzyme is located in the gastrointestinal tract where it degrades proteins to large peptides. Trypsin prefers lysine and arginine residues. One of the substrates for trypsin is chymotrypsinogen which is cleaved to produce active chymotrypsin. High levels of immunoreactive trypsin in the bloodstream can indicate pancreatic malfunction and this indicator is used to screene for and diagnose Cystic Fibrosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Anionic trypsin I; Anionic trypsin-1; Anionic trypsin-I; Beta-trypsin; Cationic trypsinogen; digestive zymogen; MGC120175; MGC149362; nonfunctional trypsin 1; pancreatic trypsin 1; Pretrypsinogen I; Protease Serine 1; protease, serine 1; protease, serine, 1; protease, serine, 1 (trypsin 1); protease, serine, 2; Serine protease 1; T-cell receptor beta chain variable region; TCR V beta 4.1; TRYP8; trypsin 1; Trypsin I; Trypsin-1; trypsinogen; trypsinogen 1; trypsinogen 16; trypsinogen A; unnamed protein product
Gene Aliases: PRSS1; PTRYI; RATPTRYI; TRP1; Try-1; TRY1; TRY4; Trygn16; TRYP1
UniProt ID: (Human) P07477, (Rat) P00762
Entrez Gene ID: (Human) 5644, (Rat) 24691, (Mouse) 114228
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support