Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
For immunocytochemistry the cells were formalin fixed.
Does not seem to have NDK kinase activity. Confers protection from cell death by Bax and alters the cellular levels of several antioxidant enzymes including Gpx5. May play a role in spermiogenesis by increasing the ability of late-stage spermatids to eliminate reactive oxygen species.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: expressed in non-metastatic cells 5; NDK-H 5; NDK5; NDP kinase homolog 5; nm23-M5; NME/NM23 family member 5; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); Nucleoside diphosphate kinase 5; nucleoside diphosphate kinase homolog 5; Putative 3'-5'-DNA exonuclease NDK5; Radial spoke head protein 23 homolog
Gene Aliases: 1700019D05Rik; Nm23-M5; Nme5; Rsph23
UniProt ID: (Mouse) Q99MH5
Entrez Gene ID: (Mouse) 75533, (Rat) 688903
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support