Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SLDAFLDLIR NLFPENLVQA CFQQIQTVTK KVLVAPPPDE EANATSAVVS LLNETVTEVP EETKMVIKKG LEFKDGMNV
Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 48%.
Excitatory amino acid transporter 2 (EAAT2) is a member of a family of solute transporter proteins. The membrane-bound protein is the principal transporter that clears the excitatory neurotransmitter glutamate from the extracellular space at synapses in the central nervous system. Glutamate clearance is necessary for proper synaptic activation and to prevent neuronal damage from excessive activation of glutamate receptors. Mutations in and decreased expression of this protein are associated with amyotrophic lateral sclerosis (ALS.) Alternatively spliced transcript variants of this gene have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAAT2; Excitatory amino acid transporter 2; excitotoxic amino acid transporter 2; GLT1a; GLT1b; GLT1c; glutamate transporter-1; Glutamate/aspartate transporter II; human brain glutamate transporter; OTTHUMP00000165209; OTTHUMP00000165210; Sodium-dependent glutamate/aspartate transporter 2; solute carrier family 1 (glial high affinity glutamate transporter), member 2; Solute carrier family 1 member 2; unnamed protein product
Gene Aliases: DEE41; EAAT2; EIEE41; GLT-1; GLT1; HBGT; SLC1A2
UniProt ID: (Human) P43004
Entrez Gene ID: (Human) 6506
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support