Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • MARK1/MARK2/MARK3 Antibodies

          Invitrogen

          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Polyclonal Antibody

          Advanced Verification
          7 Published Figures
          5 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (7)
          • Advanced Verification (1)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody (PA5-17495) in WB

          Western blot analysis of Phospho-MARK Family in extracts from Raji and BaF3 cells showing endogenous levels of phosphorylated MARK family members using Phospho-MARK Family polyclonal antibody (Product # PA5-17495). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Immunohistochemistry (IHC)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody in Western Blot (WB)
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Antibody
          Phospho-MARK1/MARK2/MARK3 (Thr215, Thr208, Thr234) Polyclonal Antibody

          Product Details

          PA5-17495

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phosphopeptide corresponding to residues surrounding threonine 215 of human MARK1
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          59 µg/mL

          Amount

          5.9 µg

          Purification

          Antigen affinity chromatography, Protein A

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10980994

          Product Specific Information

          This antibody does not react with MARK4.

          It is not recommended to aliquot this antibody.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C-TAK1; Cdc25C-associated protein kinase 1; cTAK1; ELKL motif kinase 1; ELKL motif kinase 2; ELKL motif serine-threonine protein kinase 3; ELKL motif serine/threonine-protein kinase 3; EMK-1; EMK-2; EMK1; EMK2/MARK3 serine-threonine protein kinase; MAP/microtubule affinity-regulating kinase 1; MAP/microtubule affinity-regulating kinase 2; MAP/microtubule affinity-regulating kinase 3; mark1; mark1 {ECO:0000312|RGD:619882}; mark2 {ECO:0000312|RGD:708483}; MPK-10; Par-1a; Par-1b; Par-1c; PAR1 homolog; PAR1 homolog b; PAR1 homolog c; protein kinase; Protein kinase STK10; Ser/Thr protein kinase PAR-1; Ser/Thr protein kinase PAR-1B; serine/threonine kinase; serine/threonine protein kinase EMK; Serine/threonine-protein kinase MARK1; Serine/threonine-protein kinase MARK2; Serine/threonine-protein kinase p78; testicular tissue protein Li 117; unnamed protein product

          View more View less

          Gene Aliases: 1600015G02Rik; A430080F22Rik; AW491150; B930025N23Rik; C-TAK1; CTAK1; Emk; EMK-1; EMK1; EMK2; Emk3; ETK-1; ETK1; KIAA1477; KP78; MARK; MARK1; MARK2; MARK3; mKIAA1477; mPar-1c; MPK10; PAR-1; Par-1a; Par-1b; Par-1c; PAR1A; Par1b; Par1c; VIPB

          View more View less

          UniProt ID: (Human) Q9P0L2, (Human) Q7KZI7, (Human) Q86TT8, (Mouse) Q8VHJ5, (Rat) O08678, (Rat) O08679, (Mouse) Q05512, (Rat) Q8VHF0, (Mouse) Q03141

          View more View less

          Entrez Gene ID: (Human) 4139, (Human) 2011, (Human) 4140, (Mouse) 226778, (Rat) 117016, (Rat) 60328, (Mouse) 13728, (Rat) 170577, (Mouse) 17169

          View more View less

          Function(s)
          nucleotide binding magnesium ion binding phosphatidylserine binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding phosphatidylinositol-4,5-bisphosphate binding lipid binding kinase activity transferase activity metal ion binding tau protein binding tau-protein kinase activity phosphatidic acid binding protein serine kinase activity RNA binding protein kinase activator activity cadherin binding protein phosphatase binding transferase activity, transferring phosphorus-containing groups poly(A) RNA binding non-receptor serine/threonine protein kinase
          Process(es)
          microtubule cytoskeleton organization neuron migration protein phosphorylation cytoskeleton organization positive regulation of gene expression negative regulation of gene expression negative regulation of epithelial to mesenchymal transition regulation of neuron projection development Wnt signaling pathway intracellular signal transduction regulation of dendrite development establishment of mitochondrion localization regulation of postsynapse assembly mitophagy positive regulation of neuron projection development establishment of cell polarity cell differentiation establishment or maintenance of epithelial cell apical/basal polarity protein autophosphorylation regulation of axonogenesis regulation of cytoskeleton organization mitochondrion localization axon development regulation of microtubule cytoskeleton organization establishment or maintenance of cell polarity regulating cell shape regulation of neurofibrillary tangle assembly regulation of G2/M transition of mitotic cell cycle negative regulation of hippo signaling negative regulation of protein localization to nucleus phosphorylation multicellular organism development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline