Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TGFBR3 Antibodies

          Invitrogen

          TGFBR3 Polyclonal Antibody

          Advanced Verification
          View all (20) TGFBR3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TGFBR3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          TGFBR3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TGFBR3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TGFBR3 Antibody (PA5-17529) in WB

          Western blot analysis was performed on whole cell extracts of Hep G2 (Lane 1), undifferentiated C2C12 (Lane 2), differentiated C2C12 (Lane 3), THP-1 (Lane 4), SH-SY5Y (Lane 5), Mouse Adipose (Lane 6) and Mouse Brain (Lane 7). The blot was probed with TGFBR3 Polyclonal Antibody (Product # PA5-17529, 1:1000 dilution) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjugate (Product # A27036, 0.25 µg/mL, 1:4000 dilution). A 100kDa band corresponding to TGFBR3 was observed in Hep... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TGFBR3 Antibody in Western Blot (WB)
          TGFBR3 Antibody in Western Blot (WB)
          TGFBR3 Antibody

          Product Details

          PA5-17529

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunoprecipitation (IP)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues in the extracellular domain of the type III TGF-β receptor

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          120.8 µg/mL

          Amount

          12.08 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11001447

          Product Specific Information

          It is not recommended to aliquot this antibody.

          This antibody is not cross-reactive with other family members at physiological conditions.

          Target Information

          This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Betaglycan; betaglycan proteoglycan; OTTHUMP00000011787; tgf beta receptor 3; TGF-beta receptor type 3; TGF-beta receptor type III; TGFR-3; Transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3; unnamed protein product

          View more View less

          Gene Aliases: 1110036H20Rik; AU015626; AW215636; betaglycan; BGCAN; TBRIII; TGFBR3

          View more View less

          UniProt ID: (Human) Q03167, (Mouse) O88393

          View more View less

          Entrez Gene ID: (Human) 7049, (Mouse) 21814

          View more View less

          Function(s)
          transmembrane signaling receptor activity transforming growth factor beta-activated receptor activity type II transforming growth factor beta receptor binding transforming growth factor beta receptor binding protein binding glycosaminoglycan binding heparin binding coreceptor activity fibroblast growth factor binding PDZ domain binding BMP binding SMAD binding activin binding transforming growth factor beta binding transforming growth factor beta receptor activity, type III transmembrane signal receptor
          Process(es)
          response to hypoxia epithelial to mesenchymal transition liver development heart morphogenesis muscular septum morphogenesis outflow tract morphogenesis ventricular compact myocardium morphogenesis immune response transforming growth factor beta receptor signaling pathway transforming growth factor beta receptor complex assembly positive regulation of gene expression cell migration regulation of transforming growth factor beta receptor signaling pathway negative regulation of cell migration BMP signaling pathway positive regulation of transforming growth factor beta receptor signaling pathway negative regulation of transforming growth factor beta receptor signaling pathway organ regeneration response to follicle-stimulating hormone response to prostaglandin E response to luteinizing hormone intracellular signal transduction negative regulation of I-kappaB kinase/NF-kappaB signaling regulation of JNK cascade negative regulation of epithelial cell proliferation ventricular cardiac muscle tissue morphogenesis cardiac muscle cell proliferation positive regulation of cardiac muscle cell proliferation definitive hemopoiesis cardiac epithelial to mesenchymal transition definitive erythrocyte differentiation heart trabecula formation positive regulation of SMAD protein import into nucleus negative regulation of SMAD protein import into nucleus ventricular septum morphogenesis epicardium-derived cardiac fibroblast cell development vasculogenesis involved in coronary vascular morphogenesis heart trabecula morphogenesis secondary palate development protein-containing complex assembly regulation of ERK1 and ERK2 cascade cellular response to transforming growth factor beta stimulus positive regulation of cell migration involved in sprouting angiogenesis positive regulation of transmembrane receptor protein serine/threonine kinase signaling pathway negative regulation of extracellular matrix assembly blood vessel development vasculogenesis osteoblast differentiation in utero embryonic development blastocyst development blood vessel remodeling epicardial cell to mesenchymal cell transition protein complex assembly protein phosphorylation positive regulation of cell proliferation embryo development negative regulation of epithelial cell migration negative regulation of epithelial to mesenchymal transition cell growth positive regulation of BMP signaling pathway regulation of protein binding regulation of blood vessel size positive regulation of NF-kappaB transcription factor activity negative regulation of cellular component movement palate development pathway-restricted SMAD protein phosphorylation coronary vasculature development coronary vasculature morphogenesis visceral serous pericardium development negative regulation of apoptotic process involved in morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline