Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
This antibody is predicted to react with mouse based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; BSAP; transcription factor; paired box gene 5; paired box homeotic gene 5; Paired box protein Pax-5; paired domain gene 5; Pax-5; Pax5 variant 1; Pax5 variant 2; transcription factor PAX 5
Gene Aliases: ALL3; BSAP; EBB-1; KLP; PAX-5; PAX5; RGD1565236
UniProt ID: (Human) Q02548, (Mouse) Q02650
Entrez Gene ID: (Human) 5079, (Rat) 500453, (Mouse) 18507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support