Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • LARGE Antibodies

          Invitrogen

          LARGE Polyclonal Antibody

          View all (7) LARGE antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LARGE Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          LARGE Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LARGE Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LARGE Antibody (PA5-19292) in WB

          Western blot analysis of LARGE using LARGE Polyclonal Antibody (Product # PA5-19292) (1 µg/mL) in staining of Human Kidney lysate (35 µg protein in RIPA buffer). Primary incubation was 1 hour. Detected by chemiluminescence. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LARGE Antibody in Western Blot (WB)
          LARGE Polyclonal Antibody

          Product Details

          PA5-19292

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide sequence (SEADVNSENLQKQ) corresponding to the internal amino acids of LARGE (aa 421-434).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          TBS, pH 7.3, with 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10987082

          Product Specific Information

          This antibody is predicted to react with bovine and canine based on sequence homology.

          This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

          Target Information

          The LARGE gene, which is one of the largest in the human genome, encodes a member of the N-acetylglucosaminyltransferase gene family. It encodes a glycosyltransferase which participates in glycosylation of alpha-dystroglycan, and may carry out the synthesis of glycoprotein and glycosphingolipid sugar chains. It may also be involved in the addition of a repeated disaccharide unit. Mutations in this gene cause MDC1D, a novel form of congenital muscular dystrophy with severe mental retardation and abnormal glycosylation of alpha-dystroglycan. Alternative splicing of this gene results in two transcript variants that encode the same protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Acetylglucosaminyltransferase-like 1A; acetylglucosaminyltransferase-like protein; Glycosyltransferase-like protein; glycosyltransferase-like protein LARGE1; LARGE; LARGE xylosyl- and glucuronyltransferase 1; like-acetylglucosaminyltransferase; unnamed protein product; Xylosyl- and glucuronyltransferase LARGE1

          View more View less

          Gene Aliases: KIAA0609; LARGE; LARGE1; MDC1D; MDDGA6; MDDGB6

          View more View less

          UniProt ID: (Human) O95461

          View more View less

          Entrez Gene ID: (Human) 9215

          View more View less

          Function(s)
          catalytic activity protein binding acetylglucosaminyltransferase activity glucuronosyltransferase activity transferase activity transferase activity, transferring glycosyl groups transferase activity, transferring hexosyl groups manganese ion binding UDP-xylosyltransferase activity xylosyltransferase activity metal ion binding
          Process(es)
          blood vessel development behavioral fear response neuron migration N-acetylglucosamine metabolic process protein glycosylation protein O-linked glycosylation protein targeting to membrane glycosphingolipid biosynthetic process water transport intracellular protein transport striated muscle contraction plasma membrane organization cytoskeleton organization chemical synaptic transmission neuromuscular synaptic transmission brain development heart development muscle organ development neuromuscular junction development sensory perception of sound memory protein localization determination of adult lifespan glycoprotein metabolic process glycoprotein biosynthetic process response to light stimulus response to mechanical stimulus gene expression retina layer formation myotube differentiation dentate gyrus development nerve development principal sensory nucleus of trigeminal nerve development macrophage differentiation post-embryonic hindlimb morphogenesis multicellular organism growth protein O-linked mannosylation protein modification process myelination skeletal muscle tissue regeneration post-translational protein modification muscle cell cellular homeostasis regulation of synaptic plasticity astrocyte differentiation cardiac muscle tissue development skeletal muscle fiber development synapse organization neuromuscular process controlling posture neuromuscular process synaptic growth at neuromuscular junction localization of cell striated muscle cell development cardiac muscle cell development transmembrane transport retina development in camera-type eye limb development long-term synaptic potentiation bone development skeletal muscle organ development retina vasculature development in camera-type eye connective tissue development protein-containing complex assembly basement membrane organization potassium ion transmembrane transport protein localization to membrane protein localization to plasma membrane walking behavior acetylcholine receptor signaling pathway skeletal muscle fiber differentiation reactive gliosis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline