Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
This antibody is predicted to react with baboon and rhesus monkey based on sequence homology.
Store antibody at 4°C for 1-2 weeks. For long-term storage, store at -20°C.
This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACAT; acyl coenzyme A:cholesterol acyltransferase; Acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; Carboxyesterase ES-1; carboxyesterase ES-3; Carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1-like; Carboxylesterase 1C; Carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 2 (liver); carboxylesterase; ACAT; CE-1; CEH; Cholesteryl ester hydrolase; Cocaine carboxylesterase; E1; Egasyn; ER-resident carboxylesterase; Es-22; ES-HTEL; ES-THET; ES-x; esterase 1; Esterase 2 member of carboxylesterase-cluster 1; esterase 2, member of carboxylesterase-cluster 1; esterase 22; Esterase-2; esterase-22; HMSE; human monocyte/macrophage serine esterase 1; Liver carboxylesterase 1; Liver carboxylesterase 22; liver carboxylesterase 3; Liver carboxylesterase N; Lung surfactant convertase; Methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; Neutral retinyl ester hydrolase; NREH; PES-N; pI 5.5 esterase; REH; Retinyl ester hydrolase; Serine esterase 1; TGH; triacylglycero; Triacylglycerol hydrolase; unnamed protein product
Gene Aliases: ACAT; CE-1; CEH; Ces-1; Ces-N; CES1; Ces1c; Ces1e; Ces1g; Ces1l; CES2; Ee-1; Ee1; Eg; EG244595; egasyn; Es-22; Es-4; Es-N; Es1; Es2; Es22; Es4; EsN; Gm4976; hCE-1; HMSE; HMSE1; PCE-1; PESN; REH; Ses-1; SES1; TGH
UniProt ID: (Human) P23141, (Mouse) P23953, (Rat) P10959, (Mouse) Q64176, (Mouse) Q8VCC2
Entrez Gene ID: (Human) 1066, (Rat) 679817, (Mouse) 244595, (Mouse) 13884, (Rat) 24346, (Rat) 29225, (Mouse) 13897, (Mouse) 12623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support