Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
A suggested positive control is human brain tissue lysate.
PA5-21162 can be used with blocking peptide PEP-1276.
The AP3 complex is a heterotetramer composed of two large adaptins (AP3D1, AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2). The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. AP3M1 is highly homologous to AP3M2 but is the preferred binding partner to the YQRL motif from the cytosolic domain of TGN38. AP3M1 has also been shown to interact with the HIV-1 virulence protein Nef.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 3 mu-1 subunit; adapter-related protein complex 3 subunit mu-1; adaptor related protein complex 3 mu 1 subunit; Adaptor-related protein complex 3 subunit mu-1; adaptor-related protein complex AP-3 mu 1 subunit; adaptor-related protein complex AP-3, mu 1 subunit; AP-3 adapter complex mu3A subunit; AP-3 adaptor complex mu3A subunit; AP-3 complex subunit mu-1; ap3; clathrin adaptor complex AP3, mu-3A subunit; clathrin adaptor protein mu3A; clathrin assembly protein assembly protein complex 1 medium chain homolog 1; clathrin assembly protein assembly protein complex 3 mu-1 medium chain; clathrin coat assembly protein AP47 homolog 1; clathrin coat-associated protein AP47 homolog 1; clathrin-associated adaptor protein homolog (p47A); golgi adaptor AP-1 47 kDa protein homolog 1; HA1 47 kDa subunit homolog 1; Mu-adaptin 3A; Mu3A-adaptin; unnamed protein product
Gene Aliases: 1200013D09Rik; AP3M1; C78982; P47a; R75378
UniProt ID: (Human) Q9Y2T2, (Mouse) Q9JKC8
Entrez Gene ID: (Human) 26985, (Rat) 171126, (Mouse) 55946
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support