Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, A431, HeLaS3, HepG2, Molt-4, Raji, NIH-3T3.
Predicted reactivity: Mouse (94%), Rat (94%), Bovine (94%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781P0919; DNA repair protein; DNA repair protein XRCC2; epididymis secretory sperm binding protein; recA/RAD51 family protein; unnamed protein product; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross-complementing protein 2; XRCC2
Gene Aliases: 4921524O04Rik; 8030409M04Rik; FANCU; POF17; RAD51; RecA; SPGF50; XRCC2
UniProt ID: (Human) O43543, (Mouse) Q9CX47
Entrez Gene ID: (Human) 7516, (Mouse) 57434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support