Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Recommended positive controls: 293T, HeLaS3, Raji.
Predicted reactivity: Rhesus Monkey (87%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DXYS155E is a gene found in the pseudoautosomal region of the distal short arms of the X and Y chromosomes, and appears to be ubiquitously expressed.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 385 aa isoform; 550 amino acids MW=61kDa, glycosylated=75 kDa; expressed on endothelium, activated lymphocytes and syncytiotrophoblast, contains leucine zipper and basic region homologous to myc; 721P; 695 aa isoform; 721P; A kinase (PRKA) anchor protein 17A; A-kinase anchor protein 17A; AKAP-17A; B-lymphocyte antigen; B-lymphocyte surface antigen; PRKA17A; Protein kinase A-anchoring protein 17A; Protein XE7; pseudoautosomal gene XE7; RP13-297E16.1; Splicing factor, arginine/serine-rich 17A
Gene Aliases: 721P; AKAP-17A; AKAP17A; CCDC133; CXYorf3; DXYS155E; PRKA17A; SFRS17A; XE7; XE7Y
UniProt ID: (Human) Q02040
Entrez Gene ID: (Human) 8227
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support