Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Recommended positive controls: HeLa, HCT116, A375M.
Predicted reactivity: Rhesus Monkey (99%), Bovine (83%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The WBSCR22 gene, also known as Williams-Beuren Syndrome Chromosome Region 22, encodes a protein that participates in the biogenesis of the 40S ribosomal subunit. This gene is one of the 26 deleted in Williams-Beuren syndrome, which is characterized by cognitive delays, congenital heart and vascular diseases, and distinct facial features. WBSCR22 features a nuclear localization signal and an S-adenosylmethionine (SAM) binding motif typical of methyltransferases. The protein enhances cell survival and proliferation, with increased expression linked to poor prognosis in various cancers, including glioma and breast cancer. WBSCR22 modulates key signaling pathways such as Akt/GSK3B and beta-catenin/CyclinD1, and is a direct target of miR-146b-5p. Moreover, its downregulation suppresses tumor growth, suggesting its potential as a therapeutic target in oncology.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18S rRNA (guanine-N(7))-methyltransferase; Bud site selection protein 23 homolog; Metastasis-related methyltransferase 1; methyltransferase; probable 18S rRNA (guanine-N(7))-methyltransferase; ribosome biogenesis methyltransferase WBSCR22; rRNA methyltransferase and ribosome maturation factor; unnamed protein product; Williams-Beuren candidate region putative methyltransferase; Williams-Beuren syndrome chromosomal region 22 protein; Williams-Beuren syndrome chromosomal region 22 protein homolog; Williams-Beuren syndrome chromosome region 22
Gene Aliases: 1110003N24Rik; BUD23; HASJ4442; HUSSY-03; HUSSY-3; MERM1; PP3381; WBMT; WBSCR22
UniProt ID: (Human) O43709, (Mouse) Q9CY21
Entrez Gene ID: (Human) 114049, (Mouse) 66138
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support